Anti-DES Rabbit Monoclonal Antibody

GTX61113
89363-828EA 556.26 CAD
89363-828
Anti-DES Rabbit Monoclonal Antibody
Desmin are class-III intermediate filaments found in muscle cells, and are expressed during the formation of muscle cells or fibers. Cytoskeletal intermediate filaments (IFs) are constructed from two-chain alpha-helical coiled-coil molecules arranged on an imperfect helical lattice . In adult striated muscle they form a fibrous network connecting myofibrils to each other and to the plasma membrane from the periphery of the Z-line structures. Desmin mutations have been linked to familial cardiac and skeletal myopathy .

Type: Primary
Antigen: DES
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR