Anti-DIRAS2 Rabbit Polyclonal Antibody

GTX118255
89358-866EA 258.54 CAD
89358-866
Anti-DIRAS2 Rabbit Polyclonal Antibody
DIRAS2 belongs to a distinct branch of the functionally diverse Ras (see HRAS; MIM 190020) superfamily of monomeric GTPases.[supplied by OMIM]

Type: Primary
Antigen: DIRAS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR