Anti-POLI Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX30135
89362-870EA 797.67 CAD
89362-870
Anti-POLI Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to DNA Polymerase Iota ( hRAD30B )

WB: Use at a dilution of 1/5000 (where a band is seen at ~80 kDa representing DNA Polymerase iota). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: POLI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR