Anti-Dopamine Mouse Monoclonal Antibody

Supplier: GeneTex
GTX21001
89360-936EA 639.62 CAD
89360-936
Anti-Dopamine Mouse Monoclonal Antibody
Dopamine (C6H3(OH)2-CH2-CH2-NH2) is a catecholamine neurotransmitter in the brain. Its chemical name is 4-(2-aminoethyl)benzene-1,2-diol. Dopamine is a hormone released by the hypothalamus. Its main function is to inhibit the release of prolactin from the anterior lobe of the pituitary. It can be used as a sympathomimetic drug, producing effects such as increased heart rate and blood pressure.

Type: Primary
Antigen: Dopamine
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR