Anti-DPP8 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX105200
89354-156EA 881.1 CAD
89354-156
Anti-DPP8 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to DPRP1 (dipeptidyl-peptidase 8)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: DPP8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR