Anti-DMD Mouse Monoclonal Antibody [clone: MANDYS8]

Supplier: GeneTex
GTX27163
89362-466EA 925.63 CAD
89362-466
Anti-DMD Mouse Monoclonal Antibody [clone: MANDYS8]
Antibodies
Mouse monoclonal [MANDYS8] to Dystrophin

ELISA: Use at an assay dependant dilution. ICC/IF: Use at a dilution of 1/400. IHC-Fr: Use at an assay dependant dilution. Immunohistochemical staining of normal muscle tissue results in clear labeling confined to the periphery (plasma membrane) of normal muscle fibers. WB: Use at an assay dependant dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: DMD
Clonality: Monoclonal
Clone: MANDYS8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR