Anti-E. coli Mouse Monoclonal Antibody

GTX13624
89359-780EA 378.48 CAD
89359-780
Anti-E. coli Mouse Monoclonal Antibody
Escherichia coli is a gram negative bacillus that belongs to a larger group of Enterobacteriae - bacteria that inhabit the gastrointestinal tract. Although usually a harmless resident of the gut, some strains have the potential to cause serious problems, especially where there is an immature immune system or immunosuppression, or where the subtype of organism has acquired the ability to produce pathogenic toxins.

Type: Primary
Antigen: E. coli
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR