Anti-EBV Mouse Monoclonal Antibody

GTX40449
89363-190EA 523.19 CAD
89363-190
Anti-EBV Mouse Monoclonal Antibody
Epstein Barr virus (EBV) is a member of the herpesvirus family and one of the most common human viruses. Most people become infected with EBV during their lives. Primary infections usually results in infectious mononucleosis (glandular fever) but the virus can also lay dormant in B lymphocytes and when reactivated become associated with more serious disease such as Burkitt's lymphoma, nasopharyngeal carcinoma and Hodgkin's disease.

Type: Primary
Antigen: EBV
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR