Anti-EEA1 Rabbit Monoclonal Antibody

GTX63125
89364-632EA 556.26 CAD
89364-632
Anti-EEA1 Rabbit Monoclonal Antibody
EEA1 is essential for early endosomal membrane fusion. It binds to endosomes via a C-terminal domain (EEA1-CT). It has been reported that EEA1, the Rab5 effector, is a core component of endosome docking.

Type: Primary
Antigen: EEA1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR