Anti-EGFR Rabbit Monoclonal Antibody

GTX61046
89363-784EA 556.26 CAD
89363-784
Anti-EGFR Rabbit Monoclonal Antibody
Epidermal growth factor receptor (EGFR) is a 170 kDa transmembrane glycoprotein receptor tyrosine kinase that, activated by epidermal growth factor (EGF), affects cell growth and differentiation . Binding of EGF or TGF alpha to EGFR activates tyrosine kinase activity of the receptor . Phosphorylation of Tyr 992, Tyr 1068 and Tyr 1086 is required for conformational change in the C-terminal of EGFR . Autophosphorylation of Tyr 992 creates a binding site for the phospholipase C-gamma (PLC-gamma) SH2 domain, inducing downstream signaling .

Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR