Anti-EID3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX119734
89359-062EA 881.1 CAD
89359-062
Anti-EID3 Rabbit Polyclonal Antibody
Antibodies
Act as a repressor of nuclear receptor-dependent transcription possibly by interfering with CREBBP-dependent coactivation. May function as a coinhibitor of other CREBBP/EP300-dependent transcription factors.

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: EID3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR