Anti-EMA Mouse Monoclonal Antibody [clone: EMA 201]

Supplier: GeneTex

GTX82953
89366-752EA 640.88 CAD
89366-752
Anti-EMA Mouse Monoclonal Antibody [clone: EMA 201]
Antibodies
Mouse monoclonal antibody [EMA 201] to MUC1

IHC, ICC (frozen or formalin –fixed paraffin-embedded (FFPE) tissue sections, cell smears. For IHC dilute conc. antibodies 1:50-1:200 using streptavidin~biotin system or polymer system, incubate 30 minutes at room temperature. Biotinylated antibody can be diluted 1:40-1:80 for IHC.

Type: Primary
Antigen: EMA
Clonality: Monoclonal
Clone: EMA 201
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR