Anti-EMSY Rabbit Polyclonal Antibody

GTX20123
89360-788EA 609.87 CAD
89360-788
Anti-EMSY Rabbit Polyclonal Antibody
EMSY binds BRCA2 within a region (exon 3) deleted in cancer. EMSY is capable of silencing the activation potential of BRCA2 exon 3, associates with chromatin regulators HP1 and BS69, and localizes to sites of repair following DNA damage. It's levels are amplified in breast and ovarian cancer.

Type: Primary
Antigen: EMSY
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR