Anti-Enrofloxacin/Ciprofloxacin Mouse Monoclonal Antibody

Supplier: GeneTex
GTX15060
89360-096EA 932.91 CAD
89360-096
Anti-Enrofloxacin/Ciprofloxacin Mouse Monoclonal Antibody
Antibodies
Ciprofloxacin, a fluoroquinolone, is a concentration dependent bactericidal agent. It is structurally related to enrofloxacin, and has a similar spectrum of activity. Both of these antimicrobials have shown activity against some gram positive aerobes and a wide range of gram negative bacilli and cocci, which include Klebsiella, Pseudomonas, Salmonella, and other organisms such as Mycoplasma, Staphylococci and Chlamydia. Due to the fluoroquinolones variable activity against most Streptococci, as well as their weak activity against many anaerobic bacteria, they are generally not recommended for use in treating infections where these types of microbes are present. Like enrofloxacin, ciprofloxacin is believed to act by inhibiting bacterial DNA gyrase which prevents DNA supercoiling and DNA synthesis.

Type: Primary
Antigen: Enrofloxacin/Ciprofloxacin
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR