Anti-ENTPD6 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX101851
89349-306EA 640.65 CAD
89349-306
Anti-ENTPD6 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to ENTPD6 (ectonucleoside triphosphate diphosphohydrolase 6 (putative function))

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ENTPD6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR