Anti-ERBB2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX25654
89361-684EA 920.44 CAD
89361-684
Anti-ERBB2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ErbB2 (phospho Tyr1248)

WB: Use at a concentration of 0.1 - 1.0 ?g/ml. Detects a band of approximately 185 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ERBB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR