Anti-ERG Rabbit Polyclonal Antibody

GTX124359
89359-408EA 568.8 CAD
89359-408
Anti-ERG Rabbit Polyclonal Antibody
Type: Primary
Antigen: ERG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR