Anti-Estriol Rat Monoclonal Antibody

Supplier: GeneTex
GTX72380
89353-014EA 512.31 CAD
89353-014
Anti-Estriol Rat Monoclonal Antibody
Antibodies
Estriol is the main estrogen in the blood and urine of pregnant females. Serum estriol determinations reflect the status of the fetus and are used to evaluate fetal distress and placental function.

Type: Primary
Antigen: Estriol
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR