Anti-ESR2 Rabbit Monoclonal Antibody

GTX62324
89364-402EA 556.26 CAD
89364-402
Anti-ESR2 Rabbit Monoclonal Antibody
Estrogen Receptor beta (ER beta) is a nuclear hormone receptor which is highly homologous to ER alpha. It functions as a ligand-activated transcription factor, binding to estrogens and activation expression of reporter genes containing estrogen response elements (ERE) in an estrogen-dependent manner . Studies have shown ER beta play a role in modulation of the effects of ER alpha, possibly by inhibiting the transcription activation of ER alpha . ER beta is expressed in many tissues, including human thymus, spleen, ovary and testis. It is also highly expressed in endothelial and vascular smooth muscles cells, and the expression of ER beta in these areas has a positive correlation with increased coronary artery plaque .

Type: Primary
Antigen: ESR2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR