Anti-ADGRL4 Rabbit Polyclonal Antibody

GTX108224
89355-390EA 258.54 CAD
89355-390
Anti-ADGRL4 Rabbit Polyclonal Antibody
Could be involved in cardiac development.

Type: Primary
Antigen: ADGRL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR