Anti-EZRIN Mouse Monoclonal Antibody

GTX18057
89351-436EA 529.35 CAD
89351-436
Anti-EZRIN Mouse Monoclonal Antibody
Ezrin (also known as p81, 80K, or cytovillin) is a cytoplasmic protein enriched in microvilli and other cell surface structures. Its NH2-terminal domain is associated with the plasma membrane and the COOH-terminal domain with the cytoskeleton. Ezrin has an actin-binding capability. Ezrin is expressed in epithelial cells but not in mesenchymal cells. Ezrin is also expressed by certain epithelial tumors, such as renal cell adenocarcinomas. Ezrin serves as a tyrosine kinase substrate, and is phosphorylated in EGF-stimulated cells.

Type: Primary
Antigen: EZRIN
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR