Anti-F9 Mouse Monoclonal Antibody [clone: 9D]

GTX41045
89363-424EA 443.36 CAD
89363-424
Anti-F9 Mouse Monoclonal Antibody [clone: 9D]
The vitamin K dependent coagulation factor IX circulates in the blood as an inactive zymogen at the normal concentration of approximately 3 μg/ml. This factor is converted to an active form by factor XIa, which excises the activation peptide and thus generates a heavy chain and a light chain held together by one or more disulfide bonds. The role of this activated factor IX in the blood coagulation cascade is to activate factor X to its active form through interactions with Ca+2 ions, membrane phospholipids, and factor VIII. Alterations of this gene, including point mutations, insertions and deletions, cause factor IX deficiency, which is a recessive X-linked disorder, also called hemophilia B or Christmas disease.

Type: Primary
Antigen: Factor IX
Clonality: Monoclonal
Clone: [9D]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR