Anti-F8 Mouse Monoclonal Antibody [clone: 101]

Supplier: GeneTex
GTX40721
89363-280EA 996.89 CAD
89363-280
Anti-F8 Mouse Monoclonal Antibody [clone: 101]
Antibodies
Mouse monoclonal [101] to Factor VIII

ELISA: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: F8
Clonality: Monoclonal
Clone: 101
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR