Anti-F8 Mouse Monoclonal Antibody [clone: 8A]

Supplier: GeneTex
GTX40729
89363-284EA 996.89 CAD
89363-284
Anti-F8 Mouse Monoclonal Antibody [clone: 8A]
Antibodies
Mouse monoclonal [8A] to Factor VIII

For ELISA: Use at a diution of 1:16,000. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: F8
Clonality: Monoclonal
Clone: 8A
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR