Anti-FADD Mouse Monoclonal Antibody [clone: FD19]

Supplier: Genetex
GTX10519
89354-152EA 924.88 CAD
89354-152
Anti-FADD Mouse Monoclonal Antibody [clone: FD19]
Antibodies
Mouse monoclonal [FD19] to FADD

Dot: Use at a concentration of 0.5-1 ?g/ml, determined using total cell extracts of A431 cells. ELISA: Use at an assay dependent dilution. IP: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: FADD
Clonality: Monoclonal
Clone: FD19
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR