Anti-FAS Rabbit Polyclonal Antibody

GTX15286
89351-062EA 564.78 CAD
89351-062
Anti-FAS Rabbit Polyclonal Antibody
Fas / APO1 (CD95) is an important member of the tumour necrosis factor (TNF) superfamily involved in membrane-mediated apoptosis. Ligation of Fas by Fas Ligand or an anti-Fas cross-linking antibody, triggers activation of the caspase cascade. Functional impairment of the Fas / Fas Ligand system is associated with the development and progression of malignancies. Fas gene mutations have been suggested to have a role in testicular germ cell tumours. Tumour cells frequently exhibit de novo expression of Fas Ligand, which plays a significant role in local tissue destruction, metastatic spread, and immune escape of the tumor cells. The apoptosis of lymphocytes, which occurs in autoimmune diseases, is usually induced by the Fas/Fas Ligand system. Fas is believed to be involved in various autoimmune diseases including ulcerative colitis, Graves disease, and rheumatoid arthritis. Fas expression on gastric epithelial cells in patients infected with H. Pylori is responsible for the accelerated apoptosis of the cells. Serum Fas Ligand concentration has also been shown to be associated with atherosclerosis and inflammatory disease, in patients with hypertension.

IHC-P: Use at an assay dependent dilution. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM citrate buffer, pH 6.0 for 10 min followed by cooling at RT for 20 min. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: FASLG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR