Anti-FBP1 Rabbit Monoclonal Antibody

GTX62712
89364-578EA 556.26 CAD
89364-578
Anti-FBP1 Rabbit Monoclonal Antibody
FBP1 (Fructose-1,6-bisphosphatase 1) is a gluconeogenesis regulatory enzyme that catalyzes the hydrolysis of fructose 1,6-bisphosphate to fructose 6-phosphate and inorganic phosphate . FBP1 catalyzes the removal of phosphate-1 from fructose 1,6-bisphosphate to produce fructose 6-phosphate, an important precursor in various biosynthetic pathways. It is a key, rate-controlling enzyme of gluconeogenesis, an important metabolic pathway that allows the cells to synthesize glucose and grow on non-carbohydrate carbon sources, such as glycerol, organic acids and amino acids . Fructose-1,6-diphosphatase deficiency is associated with hypoglycemia and metabolic acidosis .

Type: Primary
Antigen: FBP1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR