Anti-FBXW11 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX113940
89357-800EA 595.28 CAD
89357-800
Anti-FBXW11 Rabbit Polyclonal Antibody
Rabbit Polyclonal antibody to FBXW11 (F-box and WD repeat domain containing 11)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: FBXW11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR