Anti-FGF10 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX12469
89359-458EA 622.8 CAD
89359-458
Anti-FGF10 Rabbit Polyclonal Antibody
FGF10 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein exhibits mitogenic activity for keratinizing epidermal cells, but essentially no activity for fibroblasts, which is similar to the biological activity of FGF7. Studies of the mouse homolog of suggested that this gene is required for embryonic epidermal morphogenesis including brain development, lung morphogenesis, and initiation of lim bud formation. This gene is also implicated to be a primary factor in the process of wound healing.

ELISA: Coating concentration = 0.5 ?g/ml (100 ?l/well); Sensitivity = 0.2-0.4 ng/wellNeut: To yield one-half maximal inhibition (ND50), a 1.5 ng/ml concentration of Human FGF10 requires 0.20-0.35 ?g/ml. WB: Use at a concentration of 0.1 - 0.2 ?g/ml. Sensitivity = 1.5-3.0 ng/lane. Predicted molecular weight: 23 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: FGF10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR