Anti-FGF2 Rabbit Monoclonal Antibody

GTX62156
89364-332EA 556.26 CAD
89364-332
Anti-FGF2 Rabbit Monoclonal Antibody
Basic fibroblast growth factor (FGF2) is a wide-spectrum mitogenic, angiogenic, and neurotrophic factor that is expressed at low levels in many tissues and cell types and reaches high concentrations in brain and pituitary. FGF2 has been implicated in a multitude of physiologic and pathologic processes, including limb development, angiogenesis, wound healing, and tumor growth . It is also an important modulator of cartilage and bone growth and differentiation . FGF2 consists of multiple protein isoforms produced by alternative translation from the FGF2 gene. High molecular weight FGF2 isoforms are not secreted from the cell, but are transported to the nucleus where they regulate cell growth or behavior in an intracrine fashion. 18 kDa FGF2 can be secreted to the extracellular medium where it acts as a conventional growth factor by binding to and activation of cell-surface receptors . These protein isoforms are localized to different cellular compartments, indicating unique biological activity. FGF2 isoforms in the heart have distinct roles in many pathological circumstances in the heart including cardiac hypertrophy, ischemia-reperfusion injury, and atherosclerosis .

Type: Primary
Antigen: FGF2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR