Anti-FGA Mouse Monoclonal Antibody [clone: 15E11]

Supplier: GeneTex

GTX10063
89350-608EA 475.31 CAD
89350-608
Anti-FGA Mouse Monoclonal Antibody [clone: 15E11]
Antibodies
Mouse monoclonal [15E11] to Fibrinogen

ELISA: Use at an assay dependent dilution. (capture, detection). WB: Use at an assay dependent dilution. Use under non-reducing conditions. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: FGA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR