Anti-FLG Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX37695
89363-046EA 673.77 CAD
89363-046
Anti-FLG Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Filaggrin

For IHC-P: Use at a dilution of 1:100-1:500. For WB: Use at a dilution of 1:100-1:500. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: FLG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR