Anti-FOXA2 Mouse Monoclonal Antibody [clone: 3C10]

Supplier: GeneTex

GTX84485
89366-792EA 907.19 CAD
89366-792
Anti-FOXA2 Mouse Monoclonal Antibody [clone: 3C10]
Antibodies
Mouse monoclonal antibody [3C10] to FOXA2

Recommended Starting Dilutions:Western blot: 1:1000; IHC: 1:50; ICC/IF: 1:100; Flow 1:1,000Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: FOXA2
Clonality: Monoclonal
Clone: 3C10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR