Anti-FOXG1 Goat Polyclonal Antibody

Supplier: GeneTex
GTX23394
89361-356EA 578.99 CAD
89361-356
Anti-FOXG1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal to FOXG1

Western Blot - Approx 50-55kDa band seen in Human Brain lysate (predicted MW of 54kDa NP_004462 (FOXG1A) and NP_005240 (FOXG1B)). Recommended for use at 1.0-3.0?g/ml.

Type: Primary
Antigen: FOXG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR