Anti-FOXP1 Rabbit Monoclonal Antibody

GTX62637
89364-546EA 556.26 CAD
89364-546
Anti-FOXP1 Rabbit Monoclonal Antibody
FOXP1 belongs to subfamily P of the forkhead box (FOX) transcription factor family. Forkhead box transcription factors play important roles in the regulation of tissue- and cell type-specific transcription during both development and adulthood. FOXP1 contains protein-protein binding domains. It may act as a tumor suppressor, as it is lost in several tumor types and maps to a chromosomal region (3p14.1) reported to contain a tumor suppressor.

Type: Primary
Antigen: FOXP1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR