Anti-FSTL1 Chicken Polyclonal Antibody

GTX82279
89368-062EA 666.36 CAD
89368-062
Anti-FSTL1 Chicken Polyclonal Antibody
May modulate the action of some growth factors on cell proliferation and differentiation. Binds heparin (By similarity). This protein is a protein with similarity to follistatin. an activin-binding protein. It contains an FS module. a follistatin-like sequence containing 10 conserved cysteine residues. This protein product is thought to be an autoantigen associated with rheumatoid arthritis.

Type: Primary
Antigen: FSTL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR