Anti-GABA Guinea Pig Polyclonal Antibody

GTX31015
89367-704EA 887.22 CAD
89367-704
Anti-GABA Guinea Pig Polyclonal Antibody
Gamma-aminobutyric acid (GABA) is a major inhibitory neurotransmitter. GABA acts at inhibitory synapses in the brain and spinal cord. Inhibition is provoked by GABA binding resulting in hyperpolarization of the synaptic transmembrane potential of the affected neuron. GABA binding causes ion channels to open allowing either the flow of chloride or potassium ions into or out of the cell.

Type: Primary
Antigen: GABA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Guinea pig
Isotype: IgG
Reactivity: Other
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR