Anti-GADD45B Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX46244
89363-602EA 838.22 CAD
89363-602
Anti-GADD45B Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to GADD45B (Middle)

Recommended Dilutions: For Western Blot: Use at 0.25 ug/ml.<p>Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: GADD45B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR