Anti-GAP43 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX101535
89349-114EA 881.1 CAD
89349-114
Anti-GAP43 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to GAP43 (growth associated protein 43)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: GAP43
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR