Anti-GCDFP15 Mouse Monoclonal Antibody [clone: 23A3]

GTX17075
89351-308EA 509.54 CAD
89351-308
Anti-GCDFP15 Mouse Monoclonal Antibody [clone: 23A3]
This antibody is specific to GCDFP 15. This antibody can be used for the staining of breast carcinoma, salivary duct carcinoma and apocrine epithelia.

Type: Primary
Antigen: GCDFP15
Clonality: Monoclonal
Clone: [23A3]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR