Anti-GFAP Mouse Monoclonal Antibody [clone: GA-5]

Supplier: GeneTex

GTX29551
89352-640EA 709.31 CAD
89352-640
Anti-GFAP Mouse Monoclonal Antibody [clone: GA-5]
Antibodies
Mouse monoclonal [GA - 5] to GFAP - Astrocyte Marker prediluted

This antibody is ready to use.For best results use heat mediated antigen retrieval, this can enhance staining if tissues are over-fixed.Do not use enzymatic mediated antigen retrieval. We recommend incubating sections with the antibody for 30-60 minutes at room temperature.

Type: Primary
Antigen: GFAP
Clonality: Monoclonal
Clone: GA-5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR