Anti-GFP Mouse Monoclonal Antibody

GTX40898
89363-354EA 1036.14 CAD
89363-354
Anti-GFP Mouse Monoclonal Antibody
Type: Primary
Antigen: GFP
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Other
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR