Anti-GH1 Mouse Monoclonal Antibody [clone: Gh29]

Supplier: GeneTex
GTX21953
89351-698EA 512.1 CAD
89351-698
Anti-GH1 Mouse Monoclonal Antibody [clone: Gh29]
Antibodies
Human grwoth hormone (hGH) plays an important role in growth control. Its major role in stimulating body growth is to stimulate the liver and other tissues to secrete IGF-1. It stimulates both the differentiation and proliferation of myoblasts. It also stimulates amino acid uptake and protein synthesis in muscle and other tissues.

ELISA: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: GH1
Clonality: Monoclonal
Clone: Gh29
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR