Anti-GH1 Mouse Monoclonal Antibody [clone: GhB9]

Supplier: GeneTex
GTX21956
89351-702EA 936.72 CAD
89351-702
Anti-GH1 Mouse Monoclonal Antibody [clone: GhB9]
Antibodies
Human grwoth hormone (hGH) plays an important role in growth control. Its major role in stimulating body growth is to stimulate the liver and other tissues to secrete IGF-1. It stimulates both the differentiation and proliferation of myoblasts. It also stimulates amino acid uptake and protein synthesis in muscle and other tissues.

ELISA: Use at an assay dependent dilution. THis product can be used in an ELISA pair with GTX21954 (capture). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: GH1
Clonality: Monoclonal
Clone: GhB9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR