Anti-GJB1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX11367
89357-682EA 481.14 CAD
89357-682
Anti-GJB1 Rabbit Polyclonal Antibody
Rabbit polyclonal to Connexin 32 / GJB1

IHC-Fr: Use at a dilution of 1/400. WB: Use at a dilution of 1/400. Detects a band of approximately 27 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: GJB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR