Anti-GLI3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX104362
89350-304EA 891.75 CAD
89350-304
Anti-GLI3 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to GLI3 (GLI family zinc finger 3)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: GLI3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR