Anti-GLUT10 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX70487
89365-014EA 907.62 CAD
89365-014
Anti-GLUT10 Rabbit Polyclonal Antibody
Antibodies
Non-insulin-dependent diabetes mellitus (NIDDM) is a multifactoral disease with both environmental and genetics causes. Genome-wide screening procedures have identified several susceptibility loci for NIDDM within the human genome. A putative sugar transporter that has been localized to human chromosome 20q12-q13.1, one of the genomic loci associated with NIDDM. Because of the strong resemblance of this novel protein to members of the mammalian facilitative glucose transporter family (GLUT), the protein is known as GLUT10 (HGMW-approved gene symbol SLC2A10). Data suggests that GLUT10 an excellent candidate for a susceptibility gene involved in NIDDM.

Type: Primary
Antigen: GLUT10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR