Anti-GLUL Rabbit Polyclonal Antibody

GTX113442
89357-556EA 576.54 CAD
89357-556
Anti-GLUL Rabbit Polyclonal Antibody
Rabbit Polyclonal antibody to Glutamine synthetase (glutamate-ammonia ligase)

Suggested starting dilutions are as follows: WB: 1:500-1:20000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000, IHC-Fr: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: GLUL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Zebra Fish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR