Anti-GNMT Rabbit Polyclonal Antibody

GTX109145
89355-792EA 258.54 CAD
89355-792
Anti-GNMT Rabbit Polyclonal Antibody
The protein encoded by this gene is an enzyme that catalyzes the conversion of S-adenosyl-L-methionine (along with glycine) to S-adenosyl-L-homocysteine and sarcosine. The encoded protein is found in the cytoplasm and acts as a homotetramer. Defects in this gene are a cause of GNMT deficiency (hypermethioninemia). [provided by RefSeq]

Type: Primary
Antigen: GNMT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR