Anti-IgG Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX77194
89353-536EA 323.92 CAD
89353-536
Anti-IgG Rabbit Polyclonal Antibody
Antibodies
The affinity purified antibodies react with all Goat immunoglobulins by immunodiffusion and IEP techniques.280/495= 0.89 Molar F/P= 5.6. Rabbits were immunized with highly purified Goat IgG isolated from normal Goat serum. The resulting anti IgG was immunoaffinity purified off a Goat immunoglobulin immunoabsorbant and conjugated with fluorescein isothiocyanate (FITC).

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR